Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

551627

Sigma-Aldrich

3-Hydroxybutyronitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(OH)CH2CN
CAS Number:
Molecular Weight:
85.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.429 (lit.)

bp

214 °C (lit.)

density

0.976 g/mL at 25 °C (lit.)

SMILES string

CC(O)CC#N

InChI

1S/C4H7NO/c1-4(6)2-3-5/h4,6H,2H2,1H3

InChI key

BYJAJQGCMSBKPB-UHFFFAOYSA-N

General description

3-Hydroxybutyronitrile is a β-hydroxynitrile derivative. It undergoes thermolysis in the gas-phase through the formation of a six-membered cyclic transition state.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatenda M Mareya et al.
ChemistryOpen, 9(4), 512-520 (2020-04-30)
A comprehensive series of optimization studies including pH, solvent and temperature were completed on the nitrile hydrolyzing Rhodococcus erythropolis bacterium SET1 with the substrate 3-hydroxybutyronitrile. These identified temperature of 25 °C and pH of 7 as the best conditions to retain
Theoretical study of the thermolysis reaction of β-hydroxynitriles in the gas phase.
Chamorro E, et al.
International Journal of Quantum Chemistry, 91(5), 618-625 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service