Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

551279

Sigma-Aldrich

1-(2-Chlorophenyl)-3-methyl-2-pyrazolin-5-one

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4C3H2N2(O)CH3
CAS Number:
Molecular Weight:
208.64
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

193-197 °C (lit.)

SMILES string

CC1=NN(C(=O)C1)c2ccccc2Cl

InChI

1S/C10H9ClN2O/c1-7-6-10(14)13(12-7)9-5-3-2-4-8(9)11/h2-5H,6H2,1H3

InChI key

CWESERWNUIUBJU-UHFFFAOYSA-N

Application

1-(2-Chlorophenyl)-3-methyl-2-pyrazolin-5-one may be used in the preparation of 1-(2-chlorophenyl)-3-methylchromeno[2,3-c]pyrazol-4(1H)-one, a heterocyclic analog of xanthone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(2-Chlorophenyl)-3-methylchromeno [2, 3-c] pyrazol-4 (1H)-one.
Batezila G and Holzer W.
Molbank, 2010(1), M661-M661 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service