Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

551163

Sigma-Aldrich

Diethyl (2-cyanoethyl)phosphonate

95%

Synonym(s):

(2-Cyanoethyl)-phosphonic acid diethyl ester, 3-(Diethylphosphono)propionitrile, NSC 43781, NSC 74863

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NC(CH2)2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
191.16
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.4380 (lit.)

bp

110 °C/0.1 mmHg (lit.)

density

1.08 g/mL at 25 °C (lit.)

SMILES string

CCOP(=O)(CCC#N)OCC

InChI

1S/C7H14NO3P/c1-3-10-12(9,11-4-2)7-5-6-8/h3-5,7H2,1-2H3

InChI key

LDOZIDLMPNCNDI-UHFFFAOYSA-N

Application

Reactant for:
  • Heteroatom-directed alkyl cyanation of alkynes
  • Use as a fluorescent substrate for carbon-phosphorous lyase
  • Microwave-assisted cleavage of phosphate, phosphonate, and phorphoramide esters
  • Amination and stereoselective olefination for synthesis of HIV-1 antivirals


Reactant for synthesis of:
  • Base-catalyzed stereospecific anti-Markovnikov addition reactants
  • Lipophilic oxoamides with activity against phospholipase A2
  • Triose phosphate isomerase of muscle inhibitors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service