Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55087

Sigma-Aldrich

8-Hydroxy-2-quinolinecarbonitrile

≥98.0% (GC)

Synonym(s):

8-Hydroxyquinaldonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6N2O
CAS Number:
Molecular Weight:
170.17
Beilstein:
473250
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

functional group

nitrile

SMILES string

Oc1cccc2ccc(nc12)C#N

InChI

1S/C10H6N2O/c11-6-8-5-4-7-2-1-3-9(13)10(7)12-8/h1-5,13H

InChI key

KUQKKIBQVSFDHX-UHFFFAOYSA-N

Application

8-Hydroxy-2-quinolinecarbonitrile may be used in the preparation of 8-hydroxy-2-quinolinemethylamine, a reagent to generate peptoid oligomers.

Other Notes

Building block for preparing chelating agents

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heterocyclic amines for the construction of peptoid oligomers bearing multi-dentate ligands.
Maayan G, et al.
Tetrahedron Letters, 49(2), 335-338 (2008)
C. Moberg et al.
Journal of the Chemical Society. Chemical Communications, 810-810 (1988)
Vladimir Majerciak et al.
Journal of virology, 93(2) (2018-10-26)
Epstein-Barr virus (EBV) is a ubiquitous human pathogen associated with Burkitt's lymphoma and nasopharyngeal carcinoma. Although the EBV genome harbors more than a hundred genes, a full transcription map with EBV polyadenylation profiles remains unknown. To elucidate the 3' ends

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service