Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

550582

Sigma-Aldrich

N-Boc-3-(3-methyl-4-nitrobenzyl)-L-histidine methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26N4O6
CAS Number:
Molecular Weight:
418.44
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Boc solid-phase peptide synthesis

impurities

≤3 wt. % tert-butyl methyl ether

mp

92-104 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

COC(=O)[C@H](Cc1cn(Cc2ccc(c(C)c2)[N+]([O-])=O)cn1)NC(=O)OC(C)(C)C

InChI

1S/C20H26N4O6/c1-13-8-14(6-7-17(13)24(27)28)10-23-11-15(21-12-23)9-16(18(25)29-5)22-19(26)30-20(2,3)4/h6-8,11-12,16H,9-10H2,1-5H3,(H,22,26)/t16-/m0/s1

InChI key

QICAMETZWUJWGH-INIZCTEOSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service