Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

550515

Sigma-Aldrich

3-Oxo-1-cyclohexanecarboxylic acid

93%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H9(O)CO2H
CAS Number:
Molecular Weight:
142.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

93%

SMILES string

OC(=O)C1CCCC(=O)C1

InChI

1S/C7H10O3/c8-6-3-1-2-5(4-6)7(9)10/h5H,1-4H2,(H,9,10)

InChI key

WATQNARHYZXAGY-UHFFFAOYSA-N

General description

3-Oxo-1-cyclohexanecarboxylic acid is a cyclic keto acid that can be prepared starting from 2-acetoxy-1,4-ethoxycarbonylcyclohex-1-ne.

Application

3-Oxo-1-cyclohexanecarboxylic acid (3-oxocyclohexanecarboxylic acid) may be used in the preparation of 9-(4-chlorobenzoyl)-6-methoxy-2,3,4,9-tetrahydro-1H-carbazole-2-carboxylic acid by reacting with 4-chloro-N-(4-methoxyphenyl)benzohydrazide hydrochloride via Fischer indolization.

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2,3-Donor-Acceptor-Substituted 1,3-Butadienes. Synthesis by SO2-extrusion from 3-sulfolenes and diels-alder reactions.
Hoffmann R, et al.
Advanced Synthesis & Catalysis, 336(4), 343-349 (1994)
Development of potent and selective indomethacin analogues for the inhibition of AKR1C3 (type 5 17?-hydroxysteroid dehydrogenase/prostaglandin F synthase) in castrate-resistant prostate cancer.
Liedtke AJ, et al.
Journal of Medicinal Chemistry, 56(6), 2429-2446 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service