Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

550310

Sigma-Aldrich

6-Bromo-3-cyano-4-methylcoumarin

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3BrC4H3O2CN
CAS Number:
Molecular Weight:
264.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

186-190 °C (lit.)

SMILES string

CC1=C(C#N)C(=O)Oc2ccc(Br)cc12

InChI

1S/C11H6BrNO2/c1-6-8-4-7(12)2-3-10(8)15-11(14)9(6)5-13/h2-4H,1H3

InChI key

NPCFOYALAZMRDI-UHFFFAOYSA-N

General description

6-Bromo-3-cyano-4-methylcoumarin is a 3-cyanocoumarin derivative, which constitutes an important class of heterocycle for preparing methine dyes and bioactive compounds. It can be prepared by reacting 5′-bromo-2′-hydroxyacetophenone with malononitrile in the presence of iodine under microwave or thermal conditions.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iodine-mediated one-pot synthesis of 3-cyanocoumarins and 3-cyano-4-methylcoumarins.
Sharma D and Makrandi JK.
J. Serb. Chem. Soc., 79(5), 527-531 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service