Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

549924

Sigma-Aldrich

Glycidyl methyl ether, polymer-bound

50-90 mesh, extent of labeling: 3.0 mmol/g loading, 1 % cross-linked with divinylbenzene

Synonym(s):

Epoxide resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

crosslinking

1 % cross-linked with divinylbenzene

extent of labeling

3.0 mmol/g loading

particle size

50-90 mesh

SMILES string

C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.C(OCc3ccccc3)C4CO4

InChI

1S/C10H12O2.C10H10.C8H8/c1-2-4-9(5-3-1)6-11-7-10-8-12-10;1-3-9-5-7-10(4-2)8-6-9;1-2-8-6-4-3-5-7-8/h1-5,10H,6-8H2;3-8H,1-2H2;2-7H,1H2

InChI key

KRZGZGXXONWBRA-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service