Skip to Content
MilliporeSigma
All Photos(1)

Documents

549827

Sigma-Aldrich

4-Bromophenoxyacetic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4OCH2CO2H
CAS Number:
Molecular Weight:
231.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

impurities

4.5% 4-bromophenoxy alcohol.

mp

149-153 °C (lit.)

SMILES string

OC(=O)COc1ccc(Br)cc1

InChI

1S/C8H7BrO3/c9-6-1-3-7(4-2-6)12-5-8(10)11/h1-4H,5H2,(H,10,11)

InChI key

SZEBGAQWWSUOHT-UHFFFAOYSA-N

General description

4-Bromophenoxyacetic acid, also known as p-bromophenoxyacetate, is a phenoxyalkanoic acid derivative. It is a chemical elicitor that can induce β-glucuronidase (GUS) activity in rice.

Application

4-Bromophenoxyacetic acid may be used as a starting material in the preparation of radioiodinated phenoxyacetic acid derivatives with potential application as radiopharmaceuticals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Finding new elicitors that induce resistance in rice to the white-backed planthopper Sogatella furcifera.
He X, et al.
Bioorganic & Medicinal Chemistry Letters, 25(23), 5601-5603 (2015)
Radioiodinated phenoxyacetic acid derivatives as potential brain imaging agents. I. Efficient synthesis via trimethylsilyl intermediates.
Ohmomo Y, et al.
Chemical & Pharmaceutical Bulletin, 37(9), 2276-2281 (1989)
Chayanis Sutcharitchan et al.
Food chemistry, 322, 126766-126766 (2020-04-20)
A simple, rapid, and effective method based on salting-out extraction and LC-MS/MS techniques was developed for the determination of 39 plant growth regulator (PGR) residues in two of the most common root and rhizome Chinese herbs, Codonopsis Radix (Dangshen) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service