Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

549037

Sigma-Aldrich

4-Chlorophenyl methyl sulfone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4SO2CH3
CAS Number:
Molecular Weight:
190.65
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

95-98 °C (lit.)

SMILES string

CS(=O)(=O)c1ccc(Cl)cc1

InChI

1S/C7H7ClO2S/c1-11(9,10)7-4-2-6(8)3-5-7/h2-5H,1H3

InChI key

LMCOQDVJBWVNNI-UHFFFAOYSA-N

General description

4-Chlorophenyl methyl sulfone (CPMSO2) is a sulfone analog of 4-chlorophenyl methyl sulfide. It is used as an intermediate for preparing sulfur-containing xenobiotics.

Application

4-Chlorophenyl methyl sulfone may be used in the preparation of 4-(methylsulfonyl)aniline by reacting with ammonia. It may also be used as a standard during the quantification of 4-chlorophenyl methyl sulphide by HPLC in rat liver microsomes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New sulfonyl-containing materials for nonlinear optics: semiempirical calculations, synthesis, and properties.
Ulman A, et al.
Journal of the American Chemical Society, 112(20), 7083-7090 (1990)
Metabolism of 4-chlorophenyl methyl sulfide and its sulfone analog in cattle and sheep.
D D Oehler et al.
Archives of environmental contamination and toxicology, 12(2), 227-233 (1983-03-01)
Ivo P Nnane et al.
Journal of pharmaceutical and biomedical analysis, 27(1-2), 315-325 (2001-10-30)
Simple high performance liquid chromatography (HPLC) methods for the analysis of 4-chlorophenyl methyl sulphide (CPMS), diphenyl sulphide (DPS) and their corresponding sulphoxide and sulphone metabolites in rat liver microsomes are described. The assay methods are based on a reversed phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service