Skip to Content
MilliporeSigma
All Photos(1)

Documents

549029

Sigma-Aldrich

3-Bromo-5-fluorobenzotrifluoride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(F)CF3
CAS Number:
Molecular Weight:
243.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.456 (lit.)

bp

138-139 °C (lit.)

density

1.511 g/mL at 25 °C (lit.)

SMILES string

Fc1cc(Br)cc(c1)C(F)(F)F

InChI

1S/C7H3BrF4/c8-5-1-4(7(10,11)12)2-6(9)3-5/h1-3H

InChI key

LIGBGEJPUQBLTG-UHFFFAOYSA-N

Application

3-Bromo-5-fluorobenzotrifluoride (1-Bromo-3-fluoro-5-(trifluoromethyl)benzene) may be used in the preparation of:
  • 6-(3-fluoro-5-trifluoromethylphenyl)-hex-5-ynoic acid methyl ester
  • (E)-5-[1-(3-fluoro-5-trifluoromethylphenyl)-5-methoxycarbonyl-pent-1-enyl]-2-methoxy-3-methylbenzoic acid methyl ester
  • (E)-3-[4-(3-fluoro-5-trifluoromethylphenyl)-4-(3,4-dimethoxyphenyl)-but-3-enyl]-oxazolidin-2-one
  • (E)-5-[1-(3-fluoro-5-trifluoromethylphenyl)-4-(2-oxo-oxazolidin-3-yl)-but-1-enyl]-2-methoxy-3-methylbenzoic acid methyl ester

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, anti-HIV activity, and metabolic stability of new alkenyldiarylmethane HIV-1 non-nucleoside reverse transcriptase inhibitors.
Deng BL, et al.
Journal of Medicinal Chemistry, 48(19), 6140-6155 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service