Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

548979

Sigma-Aldrich

(1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)-1-propanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H19NO
CAS Number:
Molecular Weight:
205.30
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D −15°, c = 2 in chloroform

mp

43-47 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@H]([C@@H](O)c1ccccc1)N2CCCC2

InChI

1S/C13H19NO/c1-11(14-9-5-6-10-14)13(15)12-7-3-2-4-8-12/h2-4,7-8,11,13,15H,5-6,9-10H2,1H3/t11-,13-/m1/s1

InChI key

FZVHJGJBJLFWEX-DGCLKSJQSA-N

Application

(1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)-1-propanol can be used as:
  • A chiral catalyst to prepare corresponding enantioselective secondary aromatic alcohols by the addition of dialkylzincs to aromatic aldehydes.
  • A reactant to synthesize optically active (R)-(+)-1-phenyl-2-(1-pyrrolidinyl)-1-propanone by oxidation reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly enantioselective addition of dialkylzincs to aromatic aldehydes using 1-phenyl-2-(1-pyrrolidinyl)-1-propanol as a chiral catalyst.
Soai K, et al.
Heterocycles, 51(6), 1421-1426 (1999)
Chiral ?-alkylation/arylation in 1-phenyl-2-(1-pyrrolidinyl)-1-propanol through Grignard reactions
Kumar BNH, et al.
Tetrahedron Asymmetry, 20(24), 2773-2779 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service