Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

548723

Sigma-Aldrich

2-Bromo-4,5-dichloroimidazole

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3HBrCl2N2
CAS Number:
Molecular Weight:
215.86
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

223 °C (dec.) (lit.)

functional group

bromo
chloro

SMILES string

Clc1nc(Br)[nH]c1Cl

InChI

1S/C3HBrCl2N2/c4-3-7-1(5)2(6)8-3/h(H,7,8)

InChI key

SULQOTGFPPFUMG-UHFFFAOYSA-N

General description

2-Bromo-4,5-dichloroimidazole is a halogenated imidazole derivative that can be prepared via bromination of 4,5-dichloroimidazole using bromine.[1]

Application

2-Bromo-4,5-dichloroimidazole may be used in the synthesis of 1-methoxymethyl-2-bromo-4,5-dichloroimidazole, which shows potent insecticidal activity.[2] It may also be used to synthesize 2-substituted 4,5-dichloroimidazoles, via a novel approach that avoids the need for N-protection.[1]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insecticidal Activity of Novel Trihaloimidazole Derivatives against a Wild Colony of German Cockroaches (Blattella germanicd).
Tomioka H, et al.
Agricultural and Biological Chemistry, 55(6), 1601-1606 (1991)
Synthesis of some dihydroxyphenyl 4, 5-dichloroimidazol-2-yl ketones: compounds related to pyoluteorin.
Dirlam JP, et al.
The Journal of Organic Chemistry, 47(11), 2196-2199 (1982)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service