Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

548081

Sigma-Aldrich

2,4-Difluorobenzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3SO2Cl
CAS Number:
Molecular Weight:
212.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 548081, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

97%

refractive index

n20/D 1.518 (lit.)

bp

236-237 °C (lit.)

density

1.581 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Fc1ccc(c(F)c1)S(Cl)(=O)=O

InChI

1S/C6H3ClF2O2S/c7-12(10,11)6-2-1-4(8)3-5(6)9/h1-3H

InChI key

FJSAJUXIHJIAMD-UHFFFAOYSA-N

General description

2,4-Difluorobenzenesulfonyl chloride is an aryl sulfonyl chloride derivative.

Application

2,4-Difluorobenzenesulfonyl chloride may be used in the preparation of 2,4-difluoro-N,N-dimethylbenzenesulfonamide, a precursor for poly(aryl ether sulfonamide)s.[1]
It may be used in the preparation of the following benzenesulfonamide quinoline derivatives with potent anti-HIV-1 (human immunodeficiency virus type 1) activity:[2]
  • 2,4-difluoro-N-methyl-N-(quinolin-8-yl)benzenesulfonamide
  • N-ethyl-2,4-difluoro-N-(quinolin-8-yl)benzenesulfonamide
  • 2,4-difluoro-N-(quinolin-8-yl)benzenesulfonamide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfonamide as an Activating Group for the Synthesis of Poly (aryl ether sulfonamide) s by Nucleophilic Aromatic Substitution.
Rebeck NT and Knauss DM.
Macromolecules, 44(17), 6717-6723 (2011)
Fudi Zhong et al.
Organic & biomolecular chemistry, 13(6), 1792-1799 (2014-12-17)
Human immunodeficiency virus type 1 (HIV-1) Rev protein facilitates the export of viral RNA from nucleus to cytoplasm, which is a key step in HIV-1 pathogenesis and transmission. In this study, we have screened a commercial library and identified the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service