Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

548049

Sigma-Aldrich

Biphenyl-2-carboxaldehyde

96%

Synonym(s):

2-Biphenylcarboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CHO
CAS Number:
Molecular Weight:
182.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D >1.6220 (lit.)

bp

91-92 °C/0.15 mmHg (lit.)

density

1.130 g/mL at 25 °C (lit.)

functional group

aldehyde
phenyl

storage temp.

2-8°C

SMILES string

O=Cc1ccccc1-c2ccccc2

InChI

1S/C13H10O/c14-10-12-8-4-5-9-13(12)11-6-2-1-3-7-11/h1-10H

InChI key

LCRCBXLHWTVPEQ-UHFFFAOYSA-N

General description

Biphenyl-2-carboxaldehyde is an oxygen-containing polycyclic aromatic hydrocarbon derivative (OPAH). It can be prepared via Suzuki cross-coupling reaction between phenyl boronic acid and 2-bromobenzaldehyde.

Application

Biphenyl-2-carboxaldehyde may be used in the preparation of 9-ethylthiofluorene.

Caution

Refrigerate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis and application of ortho-palladated complex of (4-phenylbenzoylmethylene) triphenylphosphorane as a highly active catalyst in the Suzuki cross-coupling reaction.
Karami K, e tal.
Journal of Organometallic Chemistry, 696(4), 940-945 (2011)
Microbial formation and degradation of oxygen-containing polycyclic aromatic hydrocarbons (OPAHs) in soil during short-term incubation.
Wilcke W, et al.
Environmental Pollution (Barking, Essex : 1987), 184, 385-390 (2014)
The preparation of 9-alkylthiofluorenes from biphenyl-2-carboxaldehydes.
Leber JD and Elliott JD.
Tetrahedron Letters, 30(49), 6849-6850 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service