Skip to Content
MilliporeSigma
All Photos(1)

Documents

548022

Sigma-Aldrich

2,4-Dinitro-1-(trifluoromethoxy)benzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H3(NO2)2
CAS Number:
Molecular Weight:
252.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.500 (lit.)

bp

273-274 °C (lit.)

density

1.623 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(OC(F)(F)F)c(c1)[N+]([O-])=O

InChI

1S/C7H3F3N2O5/c8-7(9,10)17-6-2-1-4(11(13)14)3-5(6)12(15)16/h1-3H

InChI key

KYUUZCUXYXNPFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 4 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

349.7 °F

Flash Point(C)

176.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service