Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

547158

Sigma-Aldrich

trans,trans-2,4-Hexadienyl acetate

97%

Synonym(s):

Sorbyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH=CH)2CH2OC(O)CH3
CAS Number:
Molecular Weight:
140.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.474 (lit.)

bp

192-193 °C (lit.)

density

0.939 g/mL at 25 °C (lit.)

SMILES string

[H]/C(C)=C([H])\C([H])=C(/[H])COC(C)=O

InChI

1S/C8H12O2/c1-3-4-5-6-7-10-8(2)9/h3-6H,7H2,1-2H3/b4-3+,6-5+

InChI key

PXVKYPFROMBALG-VNKDHWASSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

181.9 °F - closed cup

Flash Point(C)

83.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service