Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

546534

Sigma-Aldrich

Tris(dimethylamido)gallium(III)

98%

Synonym(s):

Hexakis(dimethylamido)digallium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H36Ga2N6
CAS Number:
Molecular Weight:
403.90
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

98%

form

solid

reaction suitability

core: gallium
reagent type: catalyst

mp

104-105.5 °C (lit.)

SMILES string

CN(C)[Ga-]1(N(C)C)[N+](C)(C)[Ga-](N(C)C)(N(C)C)[N+]1(C)C

InChI

1S/6C2H6N.2Ga/c6*1-3-2;;/h6*1-2H3;;/q4*-1;4*+1

InChI key

RVOVCNXACQVIAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 31 Gallium

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

132.3 °F

Flash Point(C)

55.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service