Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

546453

Sigma-Aldrich

2,5-Bis(bromomethyl)-1-methoxy-4-(3′,7′-dimethyloctyloxy)benzene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(BrCH2)2C6H2(OCH3)OCH2CH2CH(CH3)(CH2)3CH(CH3)CH3
CAS Number:
Molecular Weight:
450.25
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

mp

74-77 °C (lit.)

SMILES string

COc1cc(CBr)c(OCCC(C)CCCC(C)C)cc1CBr

InChI

1S/C19H30Br2O2/c1-14(2)6-5-7-15(3)8-9-23-19-11-16(12-20)18(22-4)10-17(19)13-21/h10-11,14-15H,5-9,12-13H2,1-4H3

InChI key

DOKSAXJBKYCNRE-UHFFFAOYSA-N

Application

Conducting polymer precursor for synthesis of substituted PPV.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service