Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

545678

Sigma-Aldrich

2-Acetylthioacetophenone

97%

Synonym(s):

Phenylacyl-S-thioacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2SCOCH3
CAS Number:
Molecular Weight:
194.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

42-45 °C (lit.)

SMILES string

CC(=O)SCC(=O)c1ccccc1

InChI

1S/C10H10O2S/c1-8(11)13-7-10(12)9-5-3-2-4-6-9/h2-6H,7H2,1H3

InChI key

YHBPLTWKUZODDY-UHFFFAOYSA-N

General description

2-Acetylthioacetophenone [S-(2-oxo-2-phenylethyl) ethanethioate] is a thioacetate containing acetophenone derivative. It can be prepared from 2-bromoacetophenone and potassium thioacetate.

Application

2-Acetylthioacetophenone may be used in the preparation of 2-mercapto-1-phenylethanone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrophilic trifluoromethylthiolation of thiols with trifluoromethanesulfenamide.
Jereb M and Dolenc D.
Royal Society of Chemistry Advances, 5(72), 58292-58306 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service