Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

545392

Sigma-Aldrich

2,7-Di-tert-butyl-9-fluorenylmethanol

98%

Synonym(s):

2,7-Di-tert-butylfluorenylmethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28O
CAS Number:
Molecular Weight:
308.46
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

119-121 °C (lit.)

functional group

hydroxyl

SMILES string

CC(C)(C)c1ccc-2c(c1)C(CO)c3cc(ccc-23)C(C)(C)C

InChI

1S/C22H28O/c1-21(2,3)14-7-9-16-17-10-8-15(22(4,5)6)12-19(17)20(13-23)18(16)11-14/h7-12,20,23H,13H2,1-6H3

InChI key

HMVSJFHAWQHGEQ-UHFFFAOYSA-N

General description

2,7-Di-tert-butyl-9-fluorenylmethanol is a fluorene derivative that can be prepared from 2,7-di-tert-butylfluorene.

Application

2,7-Di-tert-butyl-9-fluorenylmethanol may be used to synthesize 2,7-di-tert-butyl-9-fluorenylmethoxycarbonyl chloride.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fmoc: a more soluble analogue of the 9-fluorenylmethoxycarbonyl protecting group.
K D Stigers et al.
The Journal of organic chemistry, 65(12), 3858-3860 (2000-06-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service