Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

544477

Sigma-Aldrich

1,6-Dimethoxyhexane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2)6OCH3
CAS Number:
Molecular Weight:
146.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.415 (lit.)

bp

182-183 °C (lit.)

density

0.851 g/mL at 25 °C (lit.)

SMILES string

COCCCCCCOC

InChI

1S/C8H18O2/c1-9-7-5-3-4-6-8-10-2/h3-8H2,1-2H3

InChI key

GZHQUHIMIVEQFV-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

156.9 °F - closed cup

Flash Point(C)

69.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael G Wade et al.
Toxicological sciences : an official journal of the Society of Toxicology, 89(1), 304-313 (2005-10-14)
Previous studies of the toxicity of candidate fuel additives identified severe testicular toxicity in animals exposed to 1,6-dimethoxyhexane (DMH). A series of studies were conducted to characterize the effects of DMH on spermatogenesis and to compare the effects of DMH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service