Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

544426

Sigma-Aldrich

4-Benzyloxybenzophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H4C(O)C6H5
CAS Number:
Molecular Weight:
288.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

solid

mp

82-85 °C (lit.)

SMILES string

O=C(c1ccccc1)c2ccc(OCc3ccccc3)cc2

InChI

1S/C20H16O2/c21-20(17-9-5-2-6-10-17)18-11-13-19(14-12-18)22-15-16-7-3-1-4-8-16/h1-14H,15H2

InChI key

NMPNTBQOLRXPGK-UHFFFAOYSA-N

General description

4-Benzyloxybenzophenone is also known as [4-(benzyloxy)phenyl](phenyl)methanone. It reacts with propiophenone in the presence of a novel titanium reagent to form triphenylbutene isomers which shows uterotropic activity.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reductive condensation of 4-benzyloxybenzophenone and propiophenone, and the estrogenic activity of 1-benzyloxy-4-(1,2-diphenylbut-1-enyl) benzene.
Zolotareva VA, et al.
ChemInform, 21(50) (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service