Skip to Content
MilliporeSigma
All Photos(1)

Documents

544256

Sigma-Aldrich

4-Ethoxycoumarin

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10O3
CAS Number:
Molecular Weight:
190.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

134-138 °C (lit.)

SMILES string

CCOC1=CC(=O)Oc2ccccc12

InChI

1S/C11H10O3/c1-2-13-10-7-11(12)14-9-6-4-3-5-8(9)10/h3-7H,2H2,1H3

InChI key

FLZDKBHAUILYAZ-UHFFFAOYSA-N

Application

4-Ethoxycoumarin may be used in the preparation of 2-substituted-6-(2-hydroxyphenyl)-4(3H)-pyrimidones. It may also be used to synthesize 2,2-diphenyl-4-chromanone and 2,2-dimethyl-4-chromanone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and some reactions of 6-(2-hydroxyphenyl)-2-thiouracils.
Morita H, et al.
Chemical & Pharmaceutical Bulletin, 31(10), 3728-3731 (1983)
The reaction of grignard reagents with 4-hydroxy-and 4-ethoxycoumarin.
Wawzonek S, et al.
Journal of the American Chemical Society, 76(4), 1080-1082 (1954)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service