Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

543519

Sigma-Aldrich

(+)-endo-2-Norborneol

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12O
CAS Number:
Molecular Weight:
112.17
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

96%

optical activity

[α]20/D +1°, c = 1 in chloroform

mp

149-154 °C (lit.)

SMILES string

O[C@@H]1CC2CCC1C2

InChI

1S/C7H12O/c8-7-4-5-1-2-6(7)3-5/h5-8H,1-4H2/t5-,6+,7-/m1/s1

InChI key

ZQTYQMYDIHMKQB-DSYKOEDSSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service