Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

543179

Sigma-Aldrich

Ethyl 2-acetoxybenzoate

98%

Synonym(s):

Ethyl acetylsalicylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(OCOCH3)CO2C2H5
CAS Number:
Molecular Weight:
208.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.506 (lit.)

bp

278-279 °C (lit.)

density

1.158 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1ccccc1OC(C)=O

InChI

1S/C11H12O4/c1-3-14-11(13)9-6-4-5-7-10(9)15-8(2)12/h4-7H,3H2,1-2H3

InChI key

UYDSGXAKLVZWIJ-UHFFFAOYSA-N

General description

Ethyl 2-acetoxybenzoate can be synthesized by reacting ethyl salicylate and acetic anhydride in the presence of zeolite Hβ. It can also be obtained from the reaction between acetylsalicyloyl chloride (in pyridine) with ethanol.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and transdermal properties of acetylsalicylic acid and selected esters
Gerber M, et al.
International Journal of Pharmaceutics, 310.1, 31-36 (2006)
Acetylation of aromatic ethers using acetic anhydride over solid acid catalysts in a solvent-free system. Scope of the reaction for substituted ethers
Smith K, et al.
Organic & Biomolecular Chemistry, 310.1, 1560-1564 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service