Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

542962

Sigma-Aldrich

4-(Trifluoromethoxy)phenylacetic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4CH2CO2H
CAS Number:
Molecular Weight:
220.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

85-88 °C (lit.)

SMILES string

OC(=O)Cc1ccc(OC(F)(F)F)cc1

InChI

1S/C9H7F3O3/c10-9(11,12)15-7-3-1-6(2-4-7)5-8(13)14/h1-4H,5H2,(H,13,14)

InChI key

ZMTIBXQPXBUWPQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian Demkowicz et al.
Chemical biology & drug design, 87(2), 233-238 (2015-08-19)
In the present work, we report the initial results of our study on a series of 3-phenylcoumarin sulfamate-based compounds containing C-F bonds as novel inhibitors of steroid sulfatase. The new compounds are potent steroid sulfatase inhibitors, possessing more than 10
Mateusz Daśko et al.
European journal of medicinal chemistry, 128, 79-87 (2017-02-06)
In the present work, we report convenient methods for the synthesis of 3-(4-aminophenyl)-coumarin-7-O-sulfamate derivatives N-acylated with fluorinated analogues of benzoic or phenylacetic acid as steroid sulfatase (STS) inhibitors. The design of these potential STS inhibitors was supported by molecular modeling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service