542792
2-Pyridylamidoxime
97%
Synonym(s):
N-Hydroxypyridine-2-carboxamidine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H7N3O
CAS Number:
Molecular Weight:
137.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
120-124 °C (lit.)
SMILES string
N\C(=N/O)c1ccccn1
InChI
1S/C6H7N3O/c7-6(9-10)5-3-1-2-4-8-5/h1-4,10H,(H2,7,9)
InChI key
XKXCGXSHUNVFCT-UHFFFAOYSA-N
Packaging
2-Pyridylamidoxime (PAO) is a pyridine derivative. It undergoes reaction with the following complexes:
- [Cr(CO)6] in THF to afford tris derivative [Cr(PAO)3]
- [Cr(CO)6] in the presence of bipyridine (bpy) to form the binuclear complex [Cr2O4(PAO)(bpy)]
- [Mo(CO)6] in air to give [Mo2O6(PAO)2]
- [Mo(CO)6] in the presence of bpy to yield the isomeric forms of [Mo(CO)4(PAO)].bpy
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Novel Chromium, Molybdenum, and Tungsten Complexes of 2-pyridylamidoxime.
Shohayeb SM, et al.
Synthesis and Reactivity in Inorganic, Metal-Organic, and Nano-Metal Chemistry, 40(10) (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service