Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

542474

Sigma-Aldrich

4-Isopropoxyphenylboronic acid

Synonym(s):

4-(1-Methylethoxy)phenylboronic acid, 4-Isopropoxybenzeneboronic acid, 4-Isopropyloxyphenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4OCH(CH3)2B(OH)2
CAS Number:
Molecular Weight:
180.01
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

SMILES string

CC(C)Oc1ccc(cc1)B(O)O

InChI

1S/C9H13BO3/c1-7(2)13-9-5-3-8(4-6-9)10(11)12/h3-7,11-12H,1-2H3

InChI key

CJUHQADBFQRIMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Contains varying amounts of anhydride

Application

Reactant for:
  • Microwave-mediated click-chemistry synthesis of glyco-porphyrin derivatives with in vitro photo-cytotoxicity for application in photodynamic therapy
  • Palladium-catalyzed oxidative cross-coupling reactions
  • Ruthenium-catalyzed hydrogenation reactions
  • Stereoselective rhodium-catalyzed arylation
  • Palladium acetate catalyzed C-glycosidation

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service