Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

542466

Sigma-Aldrich

2-Isopropoxyphenylboronic acid

Synonym(s):

2-(Propan-2-yloxy)phenylboronic acid, 2-Isopropoxybenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4OCH(CH3)2B(OH)2
CAS Number:
Molecular Weight:
180.01
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

refractive index

n20/D >1.5090 (lit.)

bp

199 °C (lit.)

mp

30-35 °C (lit.)

density

1.075 g/mL at 25 °C (lit.)

SMILES string

CC(C)Oc1ccccc1B(O)O

InChI

1S/C9H13BO3/c1-7(2)13-9-6-4-3-5-8(9)10(11)12/h3-7,11-12H,1-2H3

InChI key

XDMKBIIRBDPSOE-UHFFFAOYSA-N

Application

Reactant for:
  • Suzuki arylation
  • Palladium-catalyzed Suzuki-Miyaura reaction

Other Notes

Contains varying amounts of anhydride

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service