Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

542202

Sigma-Aldrich

2-Biphenylboronic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4B(OH)2
CAS Number:
Molecular Weight:
198.03
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

mp

167-172 °C (lit.)

functional group

phenyl

SMILES string

OB(O)c1ccccc1-c2ccccc2

InChI

1S/C12H11BO2/c14-13(15)12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9,14-15H

InChI key

HYCYKHYFIWHGEX-UHFFFAOYSA-N

Application

Reactant involved in:
  • Suzuki-Miyaura cross-coupling reactions with arylhalides, dibromovinyl precursors, alkenyl tosylates and mesylates, and quinoline carboxylates
  • Intramolecular Friedel-Crafts alkylation for synthesis of chiral tetralins
  • Hydroxylation to phenols
  • Oxidative coupling iwth alkynes
Substrate employed in a palladium-catalyzed cross-coupling with benzylic acetates providing diarylmethanes.

Other Notes

Contains varying amounts of anhydride.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryoichi Kuwano et al.
Chemical communications (Cambridge, England), (47), 5899-5901 (2005-12-01)
Benzylic acetates reacted with arylboronic acids in the presence of a DPEphos-[Pd(eta3-C3H5)Cl]2 catalyst when tert-amyl alcohol was used as a solvent, and the catalytic cross-couplings produced diarylmethanes in high yields (up to 94% isolated yield).
Chuei Wakasugi et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(24), 5449-5458 (2020-02-23)
Transition-metal complexes bearing biaryl-2,2'-diyl ligands tend to show intense luminescence. However, difficulties in synthesis have prevented their further functionalization and practical applications. Herein, a series of platinum(II) complexes bearing biaryl-2,2'-diyl ligands, which have never been prepared in air, were synthesized
Dongxue Han et al.
Nature communications, 11(1), 5659-5659 (2020-11-11)
Chiral optical materials based on circularly polarized luminescence (CPL) have emerged rapidly due to their feasible applications in diverse fields of research. However, limited to the small luminescence dissymmetry factor (glum), real application examples have rarely been reported. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service