Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

542091

Sigma-Aldrich

(S)-(−)-1-Boc-2-pyrrolidinecarbonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O2
CAS Number:
Molecular Weight:
196.25
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D −105.5°, c = 1 in chloroform

mp

33-36 °C (lit.)

SMILES string

CC(C)(C)OC(=O)N1CCC[C@H]1C#N

InChI

1S/C10H16N2O2/c1-10(2,3)14-9(13)12-6-4-5-8(12)7-11/h8H,4-6H2,1-3H3/t8-/m0/s1

InChI key

MDMSZBHMBCNYNO-QMMMGPOBSA-N

Application

Used in synthesis of prolyl oligopeptidase inhibitors.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janice Lawandi et al.
Journal of medicinal chemistry, 52(21), 6672-6684 (2009-11-06)
Prolyl oligopeptidases cleave peptides on the carboxy side of internal proline residues and their inhibition has potential in the treatment of human brain disorders. Using our docking program fitted, we have designed a series of constrained covalent inhibitors, built from
Erik A A Wallén et al.
Journal of medicinal chemistry, 46(21), 4543-4551 (2003-10-03)
Isophthalic acid bis(l-prolyl-pyrrolidine) amide is a very potent prolyl oligopeptidase inhibitor, but it has a log P value of -0.2, which is very low for a compound targeted to the brain. Therefore, these types of compounds were further modified to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service