Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

542083

Sigma-Aldrich

Lanthanum(III) acetate hydrate

≥99.99% trace metals basis

Synonym(s):

Lanthanum triacetate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
La(CH3CO2)3 · xH2O
CAS Number:
Molecular Weight:
316.04 (anhydrous basis)
Beilstein:
3693604
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥99.99% trace metals basis

reaction suitability

core: lanthanum
reagent type: catalyst

SMILES string

O.CC(=O)O[La](OC(C)=O)OC(C)=O

InChI

1S/3C2H4O2.La.H2O/c3*1-2(3)4;;/h3*1H3,(H,3,4);;1H2/q;;;+3;/p-3

InChI key

YRKCZRMEPGLHRN-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service