Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

541389

Sigma-Aldrich

2,2-Dimethyl-1,3-dioxolan-4-one

96%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$399.00

$399.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$399.00

About This Item

Empirical Formula (Hill Notation):
C5H8O3
CAS Number:
Molecular Weight:
116.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$399.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

96%

refractive index

n20/D 1.415 (lit.)

bp

41 °C (lit.)

density

0.989 g/mL at 25 °C (lit.)

functional group

ester
ether
ketal

SMILES string

CC1(C)OCC(=O)O1

InChI

1S/C5H8O3/c1-5(2)7-3-4(6)8-5/h3H2,1-2H3

InChI key

JKUCOZWILDFXMF-UHFFFAOYSA-N

General description

2,2-Dimethyl-1,3-dioxolan-4-one can be prepared via condensation reaction between glycolic acid and acetone in the presence of P2O5 in diethyl ether.[1]

Application

2,2-Dimethyl-1,3-dioxolan-4-one may be used in the synthesis of the following N,N-disubstituted amides of glycolic acid:[1]
  • N,N-diethyl-2-hydroxyacetamide
  • N,N-di(n-butyl)-2-hydroxyacetamide
  • N,N-di(n-hexyl)-2-hydroxyacetamide
  • 4-hydroxyacetylpiperidine
  • 4-hydroxyacetylmorpholine
  • N-ethyl-N-phenyl-2-hydroxyacetamide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

135.0 °F - closed cup

Flash Point(C)

57.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel method for the synthesis of 2, 2-dimethyl-1, 3-dioxolan-4-one, and its reactions with secondary amines.
Boyko VI, et al.
Synthesis, 2007(14), 2095-2096 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service