Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

540870

Sigma-Aldrich

3,5-Dichlorobenzenethiol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3SH
CAS Number:
Molecular Weight:
179.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

62-65 °C (lit.)

SMILES string

Sc1cc(Cl)cc(Cl)c1

InChI

1S/C6H4Cl2S/c7-4-1-5(8)3-6(9)2-4/h1-3,9H

InChI key

WRXIPCQPHZMXOO-UHFFFAOYSA-N

General description

3,5-Dichlorobenzenethiol (3,5-dichlorothiophenol) is a halogenated aromatic thiol.

Application

3,5-Dichlorobenzenethiol may be used to synthesize hyperbranched poly(phenylene sulfide) [HPPS] and bis-(3,5-dichlorophenyl) sulfide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of dendritic oligo (aryl sulfone) s as supports for synthesis.
Taylor PC, et al.
Tetrahedron, 61(52), 12314-12322 (2005)
Semimetallic transport in nanocomposites derived from grafting of linear and hyperbranched poly (phenylene sulfide) s onto the surface of functionalized multi-walled carbon nanotubes.
Jeon IY, et al.
Macromolecules, 41(20), 7423-7432 (2008)
Low-polydispersity Hyperbranched Polyphenylene Sulfide as a Protectant for Palladium Nanocomposites.
Meng Y, et al.
Chemistry Letters (Jpn), 44(4), 500-502 (2015)
Homan Kang et al.
Scientific reports, 5, 10144-10144 (2015-05-29)
Recently, preparation and screening of compound libraries remain one of the most challenging tasks in drug discovery, biomarker detection, and biomolecular profiling processes. So far, several distinct encoding/decoding methods such as chemical encoding, graphical encoding, and optical encoding have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service