Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

540684

Sigma-Aldrich

Methyl 2,4-dichlorobenzoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CO2CH3
CAS Number:
Molecular Weight:
205.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.557 (lit.)

bp

239 °C (lit.)

density

1.386 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1ccc(Cl)cc1Cl

InChI

1S/C8H6Cl2O2/c1-12-8(11)6-3-2-5(9)4-7(6)10/h2-4H,1H3

InChI key

VCRWILYAWSRHBN-UHFFFAOYSA-N

General description

Methyl 2,4-dichlorobenzoate is a halogenated aromatic ester. It can be synthesized from 2,4-dichlorobenzoic acid.[1]

Application

Methyl 2,4-dichlorobenzoate may be used in the synthesis of 2,4-dichloro­benzohydroxamic acid.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2, 4-Dichlorobenzohydroxamic acid.
Shang XM, et al.
Acta Crystallographica Section E, Structure Reports Online, 61(6), o1961-o1962 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service