Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

540137

Sigma-Aldrich

(2S)-(+)-Glycidyl 3-nitrobenzenesulfonate

97%

Synonym(s):

(S)-(+)-Glycidyl 3-nitrobenzenesulfonate, (S)-(+)-Oxirane-2-methanol 3-nitrobenzenesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO6S
CAS Number:
Molecular Weight:
259.24
Beilstein:
4704456
MDL number:
UNSPSC Code:
12352103

Assay

97%

optical activity

[α]20/D +22°, c = 2 in chloroform

mp

64-66 °C (lit.)

SMILES string

[O-][N+](=O)c1cccc(c1)S(=O)(=O)OC[C@@H]2CO2

Looking for similar products? Visit Product Comparison Guide

Legal Information

Manufactured under license by Rhodia Pharma Solutions, using Jacobsen HKR technology

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service