Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

539287

Sigma-Aldrich

(N,N-Dimethylaminopropyl)trimethoxysilane

96%

Synonym(s):

(Dimethylaminopropyl)trimethoxysilane, (N,N-Dimethyl-3-aminopropyl)trimethoxysilane, 3-(N,N-Dimethylamino)propyltrimethoxysilane, Dimethyl[3-(trimethoxysilyl)propyl]amine, N,N-Dimethyl-3-(trimethoxysilyl)-1-propanamine, N,N-Dimethyl-3-(trimethoxysilyl)-1-propylamine, N,N-Dimethyl-3-(trimethoxysilyl)propylamine, N,N-Dimethylaminopropyltrimethoxysilane, [3-(Dimethylamino)propyl]trimethoxysilane

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$614.00

$614.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$614.00

About This Item

Linear Formula:
(CH3)2N(CH2)3Si(OCH3)3
CAS Number:
Molecular Weight:
207.34
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$614.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

96%

form

liquid

refractive index

n20/D 1.416 (lit.)

bp

106 °C/30 mmHg (lit.)

density

0.948 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CO[Si](CCCN(C)C)(OC)OC

InChI

1S/C8H21NO3Si/c1-9(2)7-6-8-13(10-3,11-4)12-5/h6-8H2,1-5H3

InChI key

QIOYHIUHPGORLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guo Shiou Foo et al.
ChemSusChem, 10(1), 266-276 (2016-08-31)
The nature of the surface species formed through the adsorption of CO
Paul F FitzGerald et al.
Investigative radiology, 51(12), 786-796 (2016-04-27)
The aim of this study was to produce and evaluate a proposed computed tomography (CT) contrast agent based on carboxybetaine zwitterionic (CZ)-coated soluble tantalum oxide (TaO) nanoparticles (NPs). We chose tantalum to provide superior imaging performance compared with current iodine-based
Xiao-Jing Cai et al.
ACS nano, 9(8), 7940-7949 (2015-07-07)
Self-organization of nanoparticles into stable, molecularly thin films provides an insightful paradigm for manipulating the manner in which materials interact at nanoscale dimensions to generate unique material assemblies at macroscopic length scales. While prior studies in this vein have focused

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service