Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

539082

Sigma-Aldrich

Phenylsulfonylacetone

97%

Synonym(s):

Benzenesulphonylacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2CH2COCH3
CAS Number:
Molecular Weight:
198.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

55-59 °C (lit.)

SMILES string

CC(=O)CS(=O)(=O)c1ccccc1

InChI

1S/C9H10O3S/c1-8(10)7-13(11,12)9-5-3-2-4-6-9/h2-6H,7H2,1H3

InChI key

YBLGSNMIIPIRFC-UHFFFAOYSA-N

General description

Phenylsulfonylacetone is an active methylene compound. It undergoes asymmetric reduction in the presence of fermenting bakers′ yeast to afford (S)-(+)-2-hydroxypropyl phenyl suIfone.

Application

Phenylsulfonylacetone may be used in the synthesis of 2,3,5-trisubstituted furans.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative Asymmetric Reduction of Phenylthioacetone,(?)-Phenylsulfinylacetone and Phenylsulfonylacetone with Fermenting Bakers? Yeast
Iriuchijima S and Kojima N.
Agricultural and Biological Chemistry, 42.2, 451-455 (1978)
Copper-Catalyzed Coupling Cyclization of gem-Difluoroalkenes with Activated Methylene Carbonyl Compounds: Facile Domino Access to Polysubstituted Furans
Zhang, Xuxue, et al.
Organic Letters, 17.11, 2708-2711 (2015)
Arylation of enolate anions with (. eta. 6-chlorobenzene)(. eta. 5-cyclopentadienyl) iron (II) hexafluorophosphate
Moriarty RM and Gill US
Organometallics, 5.2, 253-256 (1986)
Carbanion-accelerated Claisen rearrangements
Denmark SE and Harmata MA
Journal of the American Chemical Society, 104.18, 4972-4974 (1982)
Facile three component preparation of new indolizine derivatives
Mirzaei A
The Journal of Organic Chemistry, 5.1, 971-973 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service