Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

538957

Sigma-Aldrich

Tris(benzoylacetonato) mono(phenanthroline)europium(III)

Synonym(s):

Tris(benzoylacetonato)mono(phenanthroline)europium(III)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H35EuN2O6
CAS Number:
Molecular Weight:
815.70
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

reaction suitability

reagent type: catalyst
core: europium

mp

179-186 °C (lit.)

λmax

612 nm

SMILES string

c1cnc2c(c1)ccc3cccnc23.C\C(O[Eu](O\C(C)=C\C(=O)c4ccccc4)O\C(C)=C\C(=O)c5ccccc5)=C\C(=O)c6ccccc6

InChI

1S/C12H8N2.3C10H10O2.Eu/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;3*1-8(11)7-10(12)9-5-3-2-4-6-9;/h1-8H;3*2-7,11H,1H3;/q;;;;+3/p-3/b;3*8-7+;

InChI key

CYANOTPYWCTXGU-PRTBJVMOSA-K

General description

Light emitting metal complex.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Organic Semiconductor Laser Materials

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service