Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

538426

Sigma-Aldrich

5-Chloro-2-mercaptobenzoxazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4ClNOS
CAS Number:
Molecular Weight:
185.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

280-285 °C (lit.)

SMILES string

Sc1nc2cc(Cl)ccc2o1

InChI

1S/C7H4ClNOS/c8-4-1-2-6-5(3-4)9-7(11)10-6/h1-3H,(H,9,11)

InChI key

BOBIZYYFYLLRAH-UHFFFAOYSA-N

General description

5-Chloro-2-mercaptobenzoxazole can be synthesized by reacting amino-4-chlorophenol, potassium hydroxide and carbon disulfide in ethanol.

Application

5-Chloro-2-mercaptobenzoxazole may be used in the synthesis of 5-chloro-2-(4-methyl-1-piperazinyl)benzoxazole and 5-chloro-2-morpholin-4-yl-1,3-benzoxazole.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new 5-HT3 receptor ligand. II. Structure-activity analysis of 5-HT3 receptor agonist action in the gut
Yamada M, et al.
Chemical & Pharmaceutical Bulletin, 46.3, 445-451 (1998)
A mild and efficient one-pot synthesis of 2-aminated benzoxazoles and benzothiazoles
Stewart GW, et al.
The Journal of Organic Chemistry, 74.8, 3229-3231 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service