Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

537977

Sigma-Aldrich

4-Ethyl benzenesulfonyl chloride-functionalized silica gel

200-400 mesh, extent of labeling: 1.0 mmol/g loading

Synonym(s):

Chlorosulfo-2Ar functionalized silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

extent of labeling

1.0 mmol/g loading

surface area

500 m2/g

particle size

200-400 mesh

pore size

60 Å pore size

storage temp.

2-8°C

SMILES string

O=[SiH2].CCc1ccc(cc1)S(Cl)(=O)=O

InChI

1S/C8H9ClO2S.H2OSi/c1-2-7-3-5-8(6-4-7)12(9,10)11;1-2/h3-6H,2H2,1H3;2H2

InChI key

PLUQKCYOAGXSJX-UHFFFAOYSA-N

Application

This gel can be used to immobilize alcohols and scavenge nucleophiles, as well as for “catch and release” applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service