Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

537128

Sigma-Aldrich

(S)-(−)-1-Chloro-3-(4-fluorophenoxy)-2-propanol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4OCH2CH(OH)CH2Cl
CAS Number:
Molecular Weight:
204.63
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D −0.25°, c = 1 in chloroform

refractive index

n20/D 1.522 (lit.)

bp

283-284 °C (lit.)

density

1.28 g/mL at 25 °C (lit.)

SMILES string

O[C@H](CCl)COc1ccc(F)cc1

InChI

1S/C9H10ClFO2/c10-5-8(12)6-13-9-3-1-7(11)2-4-9/h1-4,8,12H,5-6H2/t8-/m1/s1

InChI key

OSIKOYZBGUHWAB-MRVPVSSYSA-N

Legal Information

Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service