Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

536350

Sigma-Aldrich

1,8-Dinitroanthraquinone

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H6N2O6
CAS Number:
Molecular Weight:
298.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

318-322 °C (lit.)

SMILES string

[O-][N+](=O)c1cccc2C(=O)c3cccc(c3C(=O)c12)[N+]([O-])=O

InChI

1S/C14H6N2O6/c17-13-7-3-1-5-9(15(19)20)11(7)14(18)12-8(13)4-2-6-10(12)16(21)22/h1-6H

InChI key

MBIJFIUDKPXMAV-UHFFFAOYSA-N

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Ru Chen et al.
Molecules (Basel, Switzerland), 24(24) (2019-12-22)
Functional polymeric materials composed solely of lignin preparations appeared only very recently. A gradual paradigm shift spanning 56 years has revealed how lignin-lignin blends can upgrade the performance of 100 wt% lignin-based plastics. The view, first espoused in 1960, that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service