Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

535761

Sigma-Aldrich

Bis(1-octyloxy-2,2,6,6-tetramethyl-4-piperidyl) sebacate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H84N2O6
CAS Number:
Molecular Weight:
737.15
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

refractive index

n20/D 1.479 (lit.)

dynamic viscosity

3000 mPa.s(20 °C)(lit.)

transition temp

Tg >150 °C (dec)

solubility

H2O: <6 ppm at 20 °C

density

1.028 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCON1C(C)(C)CC(CC1(C)C)OC(=O)CCCCCCCCC(=O)OC2CC(C)(C)N(OCCCCCCCC)C(C)(C)C2

InChI

1S/C44H84N2O6/c1-11-13-15-17-23-27-31-49-45-41(3,4)33-37(34-42(45,5)6)51-39(47)29-25-21-19-20-22-26-30-40(48)52-38-35-43(7,8)46(44(9,10)36-38)50-32-28-24-18-16-14-12-2/h37-38H,11-36H2,1-10H3

InChI key

OSIVCXJNIBEGCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(1-octyloxy-2,2,6,6-tetramethyl-4-piperidyl) sebacate is a hindered amine light stabilizer (HALS) that is effective in retarding degradation through thermal and photo-oxidation process. It can be used in wood and plastic based coatings.

Application

Easily emulsified in water-borne systems. Miscible to >50% with xylene, MEK, and other paint solvents.
Hindered amine UV light stabilizer.
Low basicity reduces interactions with acidic paint ingredients particularly effective in coatings based on alkyds.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct detection of additives and degradation products from polymers by liquid extraction surface analysis employing chip-based nanospray mass spectrometry
Paine MRL, et al.
Rapid Communications in Mass Spectrometry, 26(4), 412-418 (2012)
Desorption electrospray ionisation mass spectrometry reveals in situ modification of a hindered amine light stabiliser resulting from direct N-OR bond cleavage
Paine MRL, et al.
Analyst, 136(5), 904-912 (2011)
Analysis of UV absorbers and stabilizers in polypropylene by liquid chromatography/atmospheric pressure chemical ionization-mass spectrometry.
Choi S and Jang J
Polymer Testing, 30(6), 673-677 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service