Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

535524

Sigma-Aldrich

5-(2-Ethoxyphenyl)-1-methyl-3-n-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]-7-pyrimidinone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4O2
CAS Number:
Molecular Weight:
312.37
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

141-145 °C (lit.)

SMILES string

CCCc1nn(C)c2C(=O)NC(=Nc12)c3ccccc3OCC

InChI

1S/C17H20N4O2/c1-4-8-12-14-15(21(3)20-12)17(22)19-16(18-14)11-9-6-7-10-13(11)23-5-2/h6-7,9-10H,4-5,8H2,1-3H3,(H,18,19,22)

InChI key

MXQUEDUMKWBYHI-UHFFFAOYSA-N

Gene Information

human ... PDE5A(8654)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Subbotina et al.
The Journal of pharmacy and pharmacology, 69(6), 675-683 (2017-02-18)
To determine the ability of 11 sildenafil analogues to discriminate between cyclic nucleotide phosphodiesterases (cnPDEs) and to characterise their inhibitory potencies (K Sildenafil analogues were identified by virtual ligand screening (VLS) and screened for their ability to inhibit adenosine cyclic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service