Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

533769

Sigma-Aldrich

2-Anilinophenylacetic acid

Synonym(s):

2-Phenylaminobenzeneacetic acid, 2-Phenylaminophenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHC6H4CH2CO2H
CAS Number:
Molecular Weight:
227.26
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

89-93 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)Cc1ccccc1Nc2ccccc2

InChI

1S/C14H13NO2/c16-14(17)10-11-6-4-5-9-13(11)15-12-7-2-1-3-8-12/h1-9,15H,10H2,(H,16,17)

InChI key

NJFCAWNKWPIBAG-UHFFFAOYSA-N

Application

  • Biologically active molecule with potential anticonvulsant properties

Reactant for:
  • Friedel-Crafts cyclization reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service