Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

533440

Sigma-Aldrich

4,5-Bis(methylthio)-1,3-dithiol-2-one

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6OS4
CAS Number:
Molecular Weight:
210.36
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

56-60 °C (lit.)

SMILES string

CSC1=C(SC)SC(=O)S1

InChI

1S/C5H6OS4/c1-7-3-4(8-2)10-5(6)9-3/h1-2H3

InChI key

ZXNVEXYJVODARP-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service