Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

532185

Sigma-Aldrich

4-Iodobenzyloxybenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4OCH2C6H5
CAS Number:
Molecular Weight:
310.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

60-63 °C (lit.)

SMILES string

Ic1ccc(OCc2ccccc2)cc1

InChI

1S/C13H11IO/c14-12-6-8-13(9-7-12)15-10-11-4-2-1-3-5-11/h1-9H,10H2

InChI key

MPWFGAWFTAZWKZ-UHFFFAOYSA-N

Application

4-Iodobenzyloxybenzene may be used in the synthesis of carborane containing mesogens.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modification of electro-optical properties of an orthoconic chiral biphenyl smectogen with its isostructural carborane analogue.
Piecek W, et al.
Journal of Materials Chemistry, 19(8), 1173-1182 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service