Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

530867

Sigma-Aldrich

Deuterium oxide

99.9 atom % D

Synonym(s):

Heavy water, Water-d2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
D2O
CAS Number:
Molecular Weight:
20.03
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

isotopic purity

99.9 atom % D

bp

101.4 °C (lit.)

mp

3.8 °C (lit.)

SMILES string

[2H]O[2H]

InChI

1S/H2O/h1H2/i/hD2

InChI key

XLYOFNOQVPJJNP-ZSJDYOACSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liset Westera et al.
Blood, 122(13), 2205-2212 (2013-08-16)
Quantitative knowledge of the turnover of different leukocyte populations is a key to our understanding of immune function in health and disease. Much progress has been made thanks to the introduction of stable isotope labeling, the state-of-the-art technique for in
Sandra J Veen et al.
Physical review letters, 109(24), 248302-248302 (2013-02-02)
By using the critical Casimir force, we study the attractive strength dependent aggregation of colloids with and without gravity by means of near field scattering. Significant differences were seen between microgravity and ground experiments, both in the structure of the
Jutta K H Skau et al.
The American journal of clinical nutrition, 101(4), 742-751 (2015-04-04)
Poor nutritional quality of complementary foods often limits growth. Animal source foods, such as milk or meat, are often unaffordable. Local affordable alternatives are needed. We evaluate the efficacy of 2 newly developed, rice-based complementary food products: WinFood (WF) with
Paul B Henrich et al.
Acta ophthalmologica, 91(2), e120-e124 (2012-11-21)
To evaluate the potential of heavier-than-water brilliant blue G (BBG-D(2) 0) to stain the internal limiting membrane (ILM) during chromovitrectomy. In a nonrandomized, prospective, clinical multicentre study, 71 consecutive chromovitrectomy interventions in 71 patients were analysed. During routine 23-gauge vitrectomy
Lars Holm et al.
American journal of physiology. Endocrinology and metabolism, 304(8), E895-E907 (2013-02-21)
A method to determine the rate of protein breakdown in individual proteins was developed and tested in rats and confirmed in humans, using administration of deuterium oxide and incorporation of the deuterium into alanine that was subsequently incorporated into body

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service